missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLCZ1 Protein
A DNA sequence encoding the Homo sapiens (Human) PLCZ1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001317698.1 |
---|---|
Gene ID (Entrez) | 89869 |
Name | PLCZ1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040761
|
enQuireBio™
QP7781-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16060761
|
enQuireBio™
QP7781-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16050761
|
enQuireBio™
QP7781-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16070761
|
enQuireBio™
QP7781-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16080761
|
enQuireBio™
QP7781-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16090761
|
enQuireBio™
QP7781-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001317698.1 | |
PLCZ1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
89869 | |
Research Use Only | |
PLCZ1 | |
Recombinant Protein | |
E. coli | |
MEMRWFLSKI QDDFRGGKIN LEKTQRLLEK LDIRCSYIHV KQIFKDNDRL KQGRITIEEF RAIYRIITHR EEIIEIFNTY SENRKILLAS NLAQFLTQEQ YAAEMSKAIA FEIIQKYEPI EEVRKAHQMS LEGFTRYMDS RECLLFKNEC RKVYQDMTHP LNDYFISSSH NTYLVSDQLL GPSDLWGYVS ALVKGCRCLE IDCWDGAQNE PVVYHGYTLT SKLLFKTVIQ AIHKYAFMTS DYPVVLSLEN HCSTAQQEVM ADNLQATFGE SLLSDMLDDF PDTLPSPEAL KFKILVKNKK IGTLKETHER KGSDKRGDNQ DKETGVKKLP GVMLFKKKKT RKLKIALALS DLVIYTKAEK FKSFQHSRLY QQFNENNSIG ETQARKLSKL RVHEFIFHTR KFITRIYPKA TRADS | |
Greater than 90% as determined by SDS-PAGE. |