missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Phosphomannomutase 2 / PMM2 / CDG1 Protein

A DNA sequence encoding the Homo sapiens (Human) Phosphomannomutase 2 / PMM2 / CDG1, was expressed in the hosts and tags indicated.

160.00€ - 2385.00€

Specifications

Accession Number NP_000294.1
Gene ID (Entrez) 5373
Name Phosphomannomutase 2 / PMM2 / CDG1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16003800
View Documents Promotion Details
enQuireBio™
QP6508-EC-100UG
100 μg
530.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
 
16023800
View Documents Promotion Details
enQuireBio™
QP6508-EC-1MG
1 mg
2385.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
 
16013800
View Documents Promotion Details
enQuireBio™
QP6508-EC-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
16033800
View Documents Promotion Details
enQuireBio™
QP6508-EC-200UG
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
16043800
View Documents Promotion Details
enQuireBio™
QP6508-EC-500UG
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
16053800
View Documents Promotion Details
enQuireBio™
QP6508-EC-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_000294.1
Phosphomannomutase 2 / PMM2 / CDG1 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5373
Research Use Only
PMM2
Recombinant Protein
E. coli
AAPGPALCLF DVDGTLTAPR QKITKEMDDF LQKLRQKIKI GVVGGSDFEK VQEQLGNDVV EKYDYVFPEN GLVAYKDGKL LCRQNIQSHL GEALIQDLIN YCLSYIAKIK LPKKRGTFIE FRNGMLNVSP IGRSCSQEER IEFYELDKKE NIRQKFVADL RKEFAGKGLT FSIGGQISFD VFPDGWDKRY CLRHVENDGY KTIYFFGDKT MPGGNDHEIF TDPRTMGYSV TAPEDTRRIC ELLFS
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications