missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PGDH / PHGDH Protein

A DNA sequence encoding the Homo sapiens (Human) PGDH / PHGDH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8438-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024621

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_006614.2
PGDH / PHGDH Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
26227
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PHGDH
Recombinant Protein
E. coli
AFANLRKVLI SDSLDPCCRK ILQDGGLQVV EKQNLSKEEL IAELQDCEGL IVRSATKVTA DVINAAEKLQ VVGRAGTGVD NVDLEAATRK GILVMNTPNG NSLSAAELTC GMIMCLARQI PQATASMKDG KWERKKFMGT ELNGKTLGIL GLGRIGREVA TRMQSFGMKT IGYDPIISPE VSASFGVQQL PLEEIWPLCD FITVHTPLLP STTGLLNDNT FAQCKKGVRV VNCARGGIVD EGALLRALQS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers