missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PFDN5 / MM-1 Protein

A DNA sequence encoding the Homo sapiens (Human) PFDN5 / MM-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8367-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16014261

  • 2385.00€ / 1mg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_002615.2
PFDN5 / MM-1 Protein
Research Use Only
PFDN5
Recombinant Protein
E. coli
AQSINITELN LPQLEMLKNQ LDQEVEFLST SIAQLKVVQT KYVEAKDCLN VLNKSNEGKE LLVPLTSSMY VPGKLHDVEH VLIDVGTGYY VEKTAEDAKD FFKRKIDFLT KQMEKIQPAL QEKHAMKQAV MEMMSQKIQQ LTALGAAQAT AKA
Greater than 90% as determined by SDS-PAGE.
5204
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers