missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Peroxisomal multifunctional enzyme type 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Peroxisomal multifunctional enzyme type 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9186-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017811

  • 645.00€ / 100µg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_000405.1
Peroxisomal multifunctional enzyme type 2 Protein
Research Use Only
HSD17B4
Recombinant Protein
Yeast
MGSPLRFDGR VVLVTGAGAG LGRAYALAFA ERGALVVVND LGGDFKGVGK GSLAADKVVE EIRRRGGKAV ANYDSVEEGE KVVKTALDAF GRIDVVVNNA GILRDRSFAR ISDEDWDIIH RVHLRGSFQV TRAAWEHMKK QKYGRIIMTS SASGIYGNFG QANYSAAKLG LLGLANSLAI EGRKSNIHCN TIAPNAGSRM TQTVMPEDLV EALKPEYVAP LVLWLCHESC EENGGLFEVG AGWIGKLRWE RTLGAIVRQK NHPMTPEAVK ANWKKICDFE NASKPQSIQE STGSIIEVLS KIDSEGGVSA NHTSRATSTA TSGFAGAIGQ KLPPFSYAYT ELEAIMYALG VGASIKDPKD LKFIYEGSSD FSCLPTFGVI IGQKSMMGGG LAEIPGLSIN FAKVLHGEQY LELYKPLPRA GKLKCEAVVA DVLDKGSGVV IIMDVYSYSE KELICHNQFS LFLVGSGGFG GKRTSDKVKV AVAIPNRPPD AVLTDTTSLN QAALYRLSGD WNPLHIDPNF ASLAGFDKPI LHGLCTFGFS ARRVLQQFAD NDVSRFKAIK ARFAKPVYPG QTLQTEMWKE GNRIHFQTKV QETGDIVISN AYVDLAPTSG TSAKTPSEGG KLQSTFVFEE IGRRLKDIGP EVVKKVNAVF EWHITKGGNI GAKWTIDLKS GSGKVYQGPA KGAADTTIIL SDEDFMEVVL GKLDPQKAFF SGRLKARGNI MLSQKLQMIL KDYAKL
Greater than 90% as determined by SDS-PAGE.
3295
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers