missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Perilipin-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Perilipin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7848-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041161

Quantity Price
1 2497.57€ / 500µg
3 2349.71€ / 500µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001073869.1
Perilipin-4 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
729359
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PLIN4
Recombinant Protein
Yeast
GVDTSKTVLT GTKDTVCSGV TGAMNVAKGT IQTGVDTTKT VLTGTKNTVC SGVTGAVNLA KEAIQGGLDT TKSMVMGTKD TMSTGLTGAA NVAKGAMQTG LNTTQNIATG T
Videos
SDS
Documents
Certificates
Special Offers

Special Offers