missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Parvalbumin / PVALB Protein

A DNA sequence encoding the Homo sapiens (Human) Parvalbumin / PVALB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6558-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16014150

  • 530.00€ / 100µg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001302461.1
Parvalbumin / PVALB Protein
Research Use Only
PVALB
Recombinant Protein
E. coli
SMTDLLNAED IKKAVGAFSA TDSFDHKKFF QMVGLKKKSA DDVKKVFHML DKDKSGFIEE DELGFILKGF SPDARDLSAK ETKMLMAAGD KDGDGKIGVD EFSTLVAES
Greater than 90% as determined by SDS-PAGE.
5816
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers