missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OTC Protein
A DNA sequence encoding the Homo sapiens (Human) OTC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6456-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000522.3 | |
OTC Protein | |
Research Use Only | |
OTC | |
Recombinant Protein | |
E. coli | |
NKVQLKGRDL LTLKNFTGEE IKYMLWLSAD LKFRIKQKGE YLPLLQGKSL GMIFEKRSTR TRLSTETGFA LLGGHPCFLT TQDIHLGVNE SLTDTARVLS SMADAVLARV YKQSDLDTLA KEASIPIING LSDLYHPIQI LADYLTLQEH YSSLKGLTLS WIGDGNNILH SIMMSAAKFG MHLQAATPKG YEPDASVTKL AEQYAKENGT KLLLTNDPLE AAHGGNVLIT DTWISMGQEE EKKKRLQAFQ GYQVTMKTAK VAASDWTFLH CLPRKPEEVD DEVFYSPRSL VFPEAENRKW TIMAVMVSLL TDYSPQLQKP KF | |
Greater than 90% as determined by SDS-PAGE. |
5009 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |