Learn More
R&D Systems™ Recombinant Human OSMR beta Protein
Product Code. 18127250
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 18127250
Brand: R&D Systems™ 4389OR050
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
The Recombinant Human OSM R beta Protein is derived from NS0. The Recombinant Human OSM R beta Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 9180
Molecular Weight (g/mol) 81 kDa
Name OSMR beta
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human OSM R beta protein Glu28-Ser739
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSM R beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.