missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolar protein 58 Protein
A DNA sequence encoding the Homo sapiens (Human) Nucleolar protein 58, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8278-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057018.1 | |
Nucleolar protein 58 Protein | |
Research Use Only | |
NOP58 | |
Recombinant Protein | |
E. coli | |
MLVLFETSVG YAIFKVLNEK KLQEVDSLWK EFETPEKANK IVKLKHFEKF QDTAEALAAF TALMEGKINK QLKKVLKKIV KEAHEPLAVA DAKLGGVIKE KLNLSCIHSP VVNELMRGIR SQMDGLIPGV EPREMAAMCL GLAHSLSRYR LKFSADKVDT MIVQAISLLD DLDKELNNYI MRCREWYGWH FPELGKIISD NLTYCKCLQK VGDRKNYASA KLSELLPEEV EAEVKAAAEI SMGTEVSEED ICNILHLCTQ VIEISEYRTQ LYEYLQNRMM AIAPNVTVMV GELVGARLIA HAGSLLNLAK HAASTVQILG AEKALFRALK SRRDTPKYGL IYHASLVGQT SPKHKGKISR MLAAKTVLAI RYDAFGEDSS SAMGVENRAK LEARLRTLED RGIRKISGTG KALAKTEKYE HKSEVKTYDP SGDSTLPTCS KKRKIEQVDK EDEITEKKAK KAKIKVKVEE EEEEKVAEEE ETSVKKKKKR GKKKHIKEEP LSEEEPCTST AIASPEKKKK KKKKRENED | |
Greater than 90% as determined by SDS-PAGE. |
51602 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |