missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nuclear transport factor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Nuclear transport factor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6442-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023340

  • 530.00€ / 100µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001308967.1
Nuclear transport factor 2 Protein
Research Use Only
NUTF2
Recombinant Protein
E. coli
MGDKPIWEQI GSSFIQHYYQ LFDNDRTQLG AIYIDASCLT WEGQQFQGKA AIVEKLSSLP FQKIQHSITA QDHQPTPDSC IISMVVGQLK ADEDPIMGFH QMFLLKNIND AWVCTNDMFR LALHNFG
Greater than 80% as determined by SDS-PAGE.
10204
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers