missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear distribution protein nudE-like 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear distribution protein nudE-like 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8111-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001020750.1 | |
Nuclear distribution protein nudE-like 1 Protein | |
Research Use Only | |
NDEL1 | |
Recombinant Protein | |
E. coli | |
MDGEDIPDFS SLKEETAYWK ELSLKYKQSF QEARDELVEF QEGSRELEAE LEAQLVQAEQ RNRDLQADNQ RLKYEVEALK EKLEHQYAQS YKQVSVLEDD LSQTRAIKEQ LHKYVRELEQ ANDDLERAKR ATIVSLEDFE QRLNQAIERN AFLESELDEK ESLLVSVQRL KDEARDLRQE LAVRERQQEV TRKSAPSSPT LDCEKMDSAV QASLSLPATP VGKGTENTFP SPKAIPNGFG TSPLTPSARI SALNIVGDLL RKVGALESKL AACRNFAKDQ ASRKSYISGN VNCGVLNGNG TKFSRSGHTS FFDKGAVNGF DTAPPPPGLG SSRPSSAPGM LPLSV | |
Greater than 80% as determined by SDS-PAGE. |
81565 | |
1 mg | |
Not determined. | |
Full Length of BC026101 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |