missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NME1 / NDKA Protein

A DNA sequence encoding the Homo sapiens (Human) NME1 / NDKA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8426-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024551

  • 2385.00€ / 1mg
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_000260.1
NME1 / NDKA Protein
Research Use Only
NME1
Recombinant Protein
E. coli
ANCERTFIAI KPDGVQRGLV GEIIKRFEQK GFRLVGLKFM QASEDLLKEH YVDLKDRPFF AGLVKYMHSG PVVAMVWEGL NVVKTGRVML GETNPADSKP GTIRGDFCIQ VGRNIIHGSD SVESAEKEIG LWFHPEELVD YTSCAQNWIY E
Greater than 90% as determined by SDS-PAGE.
4830
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers