missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Neurotrypsin Protein

A DNA sequence encoding the Homo sapiens (Human) Neurotrypsin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6537-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16094010

  • 680.00€ / 100µg

Estimated Shipment: 26-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_003610.2
Neurotrypsin Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
8492
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PRSS12
Recombinant Protein
E. coli
IIGGKNSLRG GWPWQVSLRL KSSHGDGRLL CGATLLSSCW VLTAAHCFKR YGNSTRSYAV RVGDYHTLVP EEFEEEIGVQ QIVIHREYRP DRSDYDIALV RLQGPEEQCA RFSSHVLPAC LPLWRERPQK TASNCYITGW GDTGRAYSRT LQQAAIPLLP KRFCEERYKG RFTGRMLCAG NLHEHKRVDS CQGDSGGPLM CERPGESWVV YGVTSWGYGC GVKDSPGVYT KVSAFVPWIK SVTK
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers