missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neurocalcin-delta Protein
A DNA sequence encoding the Homo sapiens (Human) Neurocalcin-delta, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001035714.1 |
---|---|
Gene ID (Entrez) | 83988 |
Name | Neurocalcin-delta Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003090
|
enQuireBio™
QP6406-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16023090
|
enQuireBio™
QP6406-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16013090
|
enQuireBio™
QP6406-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16033090
|
enQuireBio™
QP6406-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16043090
|
enQuireBio™
QP6406-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053090
|
enQuireBio™
QP6406-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001035714.1 | |
Neurocalcin-delta Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
83988 | |
Research Use Only | |
NCALD | |
Recombinant Protein | |
E. coli | |
GKQNSKLRPE VMQDLLESTD FTEHEIQEWY KGFLRDCPSG HLSMEEFKKI YGNFFPYGDA SKFAEHVFRT FDANGDGTID FREFIIALSV TSRGKLEQKL KWAFSMYDLD GNGYISKAEM LEIVQAIYKM VSSVMKMPED ESTPEKRTEK IFRQMDTNRD GKLSLEEFIR GAKSDPSIVR LLQCDPSSAG QF | |
Greater than 80% as determined by SDS-PAGE. |