missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NEDD8 Protein

A DNA sequence encoding the Homo sapiens (Human) NEDD8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7554-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029350

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_006147.1
NEDD8 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
4738
Greater than 80% as determined by SDS-PAGE.
Research Use Only
NEDD8
Recombinant Protein
E. coli
MLIKVKTLTG KEIEIDIEPT DKVERIKERV EEKEGIPPQQ QRLIYSGKQM NDEKTAADYK ILGGSVLHLV LALRGG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers