missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MSRB3 Protein
A DNA sequence encoding the Homo sapiens (Human) MSRB3, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001026849.1 |
---|---|
Gene ID (Entrez) | 253827 |
Name | MSRB3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010941
|
enQuireBio™
QP7810-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16030941
|
enQuireBio™
QP7810-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16020941
|
enQuireBio™
QP7810-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16040941
|
enQuireBio™
QP7810-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16050941
|
enQuireBio™
QP7810-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16060941
|
enQuireBio™
QP7810-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001026849.1 | |
MSRB3 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
253827 | |
Research Use Only | |
MSRB3 | |
Recombinant Protein | |
E. coli | |
MSAFNLLHLV TKSQPVALRA CGLPSGSCRD KKNCKVVFSQ QELRKRLTPL QYHVTQEKGT ESAFEGEYTH HKDPGIYKCV VCGTPLFKSE TKFDSGSGWP SFHDVINSEA ITFTDDFSYG MHRVETSCSQ CGAHLGHIFD DGPRPTGKRY CINSAALSFT PADSSGTAEG GSGVASPAQA DKAEL | |
Greater than 80% as determined by SDS-PAGE. |