missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MORF4 family-associated protein 1-like 1 Protein

A DNA sequence encoding the Homo sapiens (Human) MORF4 family-associated protein 1-like 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6379-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022890

  • 645.00€ / 100µg
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_982287.1
MORF4 family-associated protein 1-like 1 Protein
Research Use Only
MRFAP1L1
Recombinant Protein
Yeast
MRPLDIDEVE APEEVEVLEP EEDFEQFLLP VINEMREDIA SLIREHGRAY LRTRSKLWEM DNMLIQIKTQ VEASEESALN HVQHPSGEAD ERVSELCEKA EEKAKEIAKM AEMLVELVWR IERSESS
Greater than 90% as determined by SDS-PAGE.
114932
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers