missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Modulator of apoptosis 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Modulator of apoptosis 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6372-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042830

  • 2248.90€ / 1mg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_071434.2
Modulator of apoptosis 1 Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
64112
Greater than 80% as determined by SDS-PAGE.
Research Use Only
MOAP1
Recombinant Protein
E. coli
MTLRLLEDWC RGMDMNPRKA LLIAGISQSC SVAEIEEALQ AGLAPLGEYR LLGRMFRRDE NRKVALVGLT AETSHALVPK EIPGKGGIWR VIFKPPDPDN TFLSRLNEFL AGEGMTVGEL SRALGHENGS LDPEQGMIPE MWAPMLAQAL EALQPALQCL KYKKLRVFSG RESPEPGEEE FGRWMFHTTQ MIKAWQVPDV EKRRRLLESL RGPALDVIRV LKINNPLITV DECLQALEEV FGVTDNPREL QVKYLTTYQK DEEKLSAYVL RLEPLLQKLV QRGAIERDAV NQARLDQVIA GAVHKTIRRE LNLPEDGPAP GFLQLLVLIK DYEAAEEEEA LLQAILEGNF T
Videos
SDS
Documents
Certificates
Special Offers

Special Offers