missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MKK6 / MEK6 / MAP2K6 Protein
A DNA sequence encoding the Homo sapiens (Human) MKK6 / MEK6 / MAP2K6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6338-ba-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002749.2 | |
MKK6 / MEK6 / MAP2K6 Protein | |
Research Use Only | |
MAP2K6 | |
Recombinant Protein | |
Baculovirus | |
MSQSKGKKRN PGLKIPKEAF EQPQTSSTPP RDLDSKACIS IGNQNFEVKA DDLEPIMELG RGAYGVVEKM RHVPSGQIMA VKRIRATVNS QEQKRLLMDL DISMRTVDCP FTVTFYGALF REGDVWICME LMDTSLDKFY KQVIDKGQTI PEDILGKIAV SIVKALEHLH SKLSVIHRDV KPSNVLINAL GQVKMCDFGI SGYLVDSVAK TIDAGCKPYM APERINPELN QKGYSVKSDI WSLGITMIEL AILRFPYDSW GTPFQQLKQV VEEPSPQLPA DKFSAEFVDF TSQCLKKNSK ERPTYPELMQ HPFFTLHESK GTDVASFVKL ILGD | |
Greater than 90% as determined by SDS-PAGE. |
5608 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |