missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Minor capsid protein L2 Protein
A DNA sequence encoding the Homo sapiens (Human) Minor capsid protein L2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7293-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_040316.1 | |
Minor capsid protein L2 Protein | |
Research Use Only | |
L2 | |
Recombinant Protein | |
Yeast | |
MVSHRAARRK RASVTDLYKT CKQSGTCPPD VVPKVEGTTL ADKILQWSSL GIFLGGLGIG TGSGTGGRTG YIPLGGRSNT VVDVGPTRPP VVIEPVGPTD PSIVTLIEDS SVVTSGAPRP TFTGTSGFDI TSAGTTTPAV LDITPSSTSV SISTTNFTNP AFSDPSIIEV PQTGEVAGNV FVGTPTSGTH GYEEIPLQTF ASSGTGEEPI SSTPLPTVRR VAGPRLYSRA YQQVSVANPE FLTRPSSLIT YDNPAFEPVD TTLTFDPRSD VPDSDFMDII RLHRPALTSR RGTVRFSRLG QRATMFTRSG TQIGARVHFY HDISPIAPSP EYIELQPLVS ATEDNDLFDI YADDMDPAVP VPSRSTTSFA FFKYSPTISS ASSYSNVTVP LTSSWDVPVY TGPDITLPST TSVWPIVSPT APASTQYIGI HGTHYYLWPL YYFIPKKRKR VPYFFADGFV AA | |
Greater than 90% as determined by SDS-PAGE. |
1489091 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |