missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MFG-E8 / lactadherin Protein
A DNA sequence encoding the Homo sapiens (Human) MFG-E8 / lactadherin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6356-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001108086.1 | |
MFG-E8 / lactadherin Protein | |
Research Use Only | |
MFGE8 | |
Recombinant Protein | |
E. coli | |
LDICSKNPCH NGGLCEEISQ EVRGDVFPSY TCTCLKGYAG NHCETKCVEP LGMENGNIAN SQIAASSVRV TFLGLQHWVP ELARLNRAGM VNAWTPSSND DNPWIQVNLL RRMWVTGVVT QGASRLASHE YLKAFKVAYS LNGHEFDFIH DVNKKHKEFV GNWNKNAVHV NLFETPVEAQ YVRLYPTSCH TACTLRFELL GCELNGCANP LGLKNNSIPD KQITASSSYK TWGLHLFSWN PSYARLDKQG NFNAWVAGSY GNDQWLQVDL GSSKEVTGII TQGARNFGSV QFVASYKVAY SNDSANWTEY QDPRTGSSKI FPGNWDNHSH KKNLFETPIL ARYVRILPVA WHNRIALRLE LLGC | |
Greater than 90% as determined by SDS-PAGE. |
4240 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |