missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Meteorin-like Protein

A DNA sequence encoding the Homo sapiens (Human) Meteorin-like protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7719-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020401

Quantity Price
1 1600.18€ / 100µg
3 1504.97€ / 100µg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001004431.1
Meteorin-like Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
284207
Greater than 90% as determined by SDS-PAGE.
Research Use Only
METRNL
Recombinant Protein
Mammal
QYSSDRCSWK GSGLTHEAHR KEVEQVYLRC AAGAVEWMYP TGALIVNLRP NTFSPARHLT VCIRSFTDSS GANIYLEKTG ELRLLVPDGD GRPGRVQCFG LEQGGLFVEA TPQQDIGRRT TGFQYELVRR HRASDLHELS APCRPCSDTE VLLAVCTSDF AVRGSIQQVT HEPERQDSAI HLRVSRLYRQ KSRVFEPVPE GDGHWQGRVR TLLECGVRPG HGDFLFTGHM HFGEARLGCA PRFKDFQRMY RDAQERGLNP CEVGTD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers