missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lymphocyte antigen 6H Protein
A DNA sequence encoding the Homo sapiens (Human) Lymphocyte antigen 6H, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9259-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001123950.1 | |
Lymphocyte antigen 6H Protein | |
Research Use Only | |
LY6H | |
Recombinant Protein | |
Yeast | |
LWCQDCTLTT NSSHCTPKQC QPSDTVCASV RITDPSSSRK DHSVNKMCAS SCDFVKRHFF SDYLMGFINS GILKVDVDCC EKDLCNGAAG | |
Greater than 90% as determined by SDS-PAGE. |
4062 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |