missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LY6G6D Protein

A DNA sequence encoding the Homo sapiens (Human) LY6G6D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9258-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027931

Quantity Price
1 595.74€ / 100µg
5 560.65€ / 100µg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_067069.2
LY6G6D Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
58530
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LY6G6D
Recombinant Protein
Yeast
NRMRCYNCGG SPSSSCKEAV TTCGEGRPQP GLEQIKLPGN PPVTLIHQHP ACVAAHHCNQ VETESVGDVT YPAHRDCYLG DLCNS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers