missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LY6G6D Protein
A DNA sequence encoding the Homo sapiens (Human) LY6G6D, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_067069.2 |
---|---|
Gene ID (Entrez) | 58530 |
Name | LY6G6D Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027931
|
enQuireBio™
QP9258-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16067931
|
enQuireBio™
QP9258-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16037931
|
enQuireBio™
QP9258-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16057931
|
enQuireBio™
QP9258-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16077931
|
enQuireBio™
QP9258-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16047931
|
enQuireBio™
QP9258-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_067069.2 | |
LY6G6D Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
58530 | |
Research Use Only | |
LY6G6D | |
Recombinant Protein | |
Yeast | |
NRMRCYNCGG SPSSSCKEAV TTCGEGRPQP GLEQIKLPGN PPVTLIHQHP ACVAAHHCNQ VETESVGDVT YPAHRDCYLG DLCNS | |
Greater than 90% as determined by SDS-PAGE. |