missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Luc7-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Luc7-like protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7504-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029050

  • 530.00€ / 100µg
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_006098.2
Luc7-like protein 1
Research Use Only
LUC7L3
Recombinant Protein
E. coli
MISAAQLLDE LMGRDRNLAP DEKRSNVRWD HESVCKYYLC GFCPAELFTN TRSDLGPCEK IHDENLRKQY EKSSRFMKV
Greater than 90% as determined by SDS-PAGE.
51747
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers