missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Keratin, type II cytoskeletal 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Keratin, type II cytoskeletal 4, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Name | Keratin, type II cytoskeletal 4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | KRT4 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16032160
|
enQuireBio™
QP6281-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16072160
|
enQuireBio™
QP6281-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16042160
|
enQuireBio™
QP6281-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062160
|
enQuireBio™
QP6281-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16082160
|
enQuireBio™
QP6281-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052160
|
enQuireBio™
QP6281-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Keratin, type II cytoskeletal 4 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
KRT4 | |
Recombinant Protein | |
Yeast | |
MIARQQCVRG GPRGFSCGSA IVGGGKRGAF SSVSMSGGAG RCSSGGFGSR SLYNLRGNKS ISMSVAGSRQ GACFGGAGGF GTGGFGAGGF GAGFGTGGFG GGFGGSFSGK GGPGFPVCPA GGIQEVTINQ SLLTPLHVEI DPEIQKVRTE EREQIKLLNN KFASFIDKVQ FLEQQNKVLE TKWNLLQQQT TTTSSKNLEP LFETYLSVLR KQLDTLGNDK GRLQSELKTM QDSVEDFKTK YEEEINKRTA AENDFVVLKK DVDAAYLNKV ELEAKVDSLN DEINFLKVLY DAELSQMQTH VSDTSVVLSM DNNRNLDLDS IIAEVRAQYE EIAQRSKAEA EALYQTKVQQ LQISVDQHGD NLKNTKSEIA ELNRMIQRLR AEIENIKKQC QTLQVSVADA EQRGENALKD AHSKRVELEA ALQQAKEELA RMLREYQELM SVKLALDIEI ATYRKLLEGE EYRMSGECQS AVSISVVSGS TSTGGISGGL GSGSGFGLSS GFGSGSGSGF GFGGSVSGSS SSKIISTTTL NKRR | |
Greater than 90% as determined by SDS-PAGE. |