missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCNJ10 Protein
A DNA sequence encoding the Homo sapiens (Human) KCNJ10, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_002232.2 |
---|---|
Gene ID (Entrez) | 3766 |
Name | KCNJ10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011970
|
enQuireBio™
QP6256-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16031970
|
enQuireBio™
QP6256-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16021970
|
enQuireBio™
QP6256-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041970
|
enQuireBio™
QP6256-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16051970
|
enQuireBio™
QP6256-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061970
|
enQuireBio™
QP6256-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_002232.2 | |
KCNJ10 Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3766 | |
Research Use Only | |
KCNJ10 | |
Recombinant Protein | |
E. coli | |
FLAKIARPKK RAETIRFSQH AVVASHNGKP CLMIRVANMR KSLLIGCQVT GKLLQTHQTK EGENIRLNQV NVTFQVDTAS DSPFLILPLT FYHVVDETSP LKDLPLRSGE GDFELVLILS GTVESTSATC QVRTSYLPEE ILWGYEFTPA ISLSASGKYI ADFSLFDQVV KVASPSGLRD STVRYGDPEK LKLEESLREQ AEKEGSALSV RISNV | |
Greater than 90% as determined by SDS-PAGE. |