missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCNJ10 Protein
A DNA sequence encoding the Homo sapiens (Human) KCNJ10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6256-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002232.2 | |
KCNJ10 Protein | |
Research Use Only | |
KCNJ10 | |
Recombinant Protein | |
E. coli | |
FLAKIARPKK RAETIRFSQH AVVASHNGKP CLMIRVANMR KSLLIGCQVT GKLLQTHQTK EGENIRLNQV NVTFQVDTAS DSPFLILPLT FYHVVDETSP LKDLPLRSGE GDFELVLILS GTVESTSATC QVRTSYLPEE ILWGYEFTPA ISLSASGKYI ADFSLFDQVV KVASPSGLRD STVRYGDPEK LKLEESLREQ AEKEGSALSV RISNV | |
Greater than 90% as determined by SDS-PAGE. |
3766 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |