missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IP-10 / CXCL10 Protein

A DNA sequence encoding the Homo sapiens (Human) IP-10 / CXCL10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8532-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015141

  • 645.00€ / 100µg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001556.2
IP-10 / CXCL10 Protein
Research Use Only
CXCL10
Recombinant Protein
Yeast
VPLSRTVRCT CISISNQPVN PRSLEKLEII PASQFCPRVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP
Greater than 90% as determined by SDS-PAGE.
3627
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers