missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Integrin alpha-V Protein

A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-V, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9224-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017871

Quantity Price
1 595.74€ / 100µg
5 560.65€ / 100µg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001138471.1
Integrin alpha-V Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
3685
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ITGAV
Recombinant Protein
Yeast
DLALSEGDIH TLGCGVAQCL KIVCQVGRLD RGKSAILYVK SLLWTETFMN KENQNHSYSL KSSASFNVIE FPYKNLPIED ITNSTLVTTN VTWGIQPAPM PVPVWVIILA VLAGLLLLAV LVFVMYRMGF FKRVRPPQEE QEREQLQPHE NGEGNSET
Videos
SDS
Documents
Certificates
Special Offers

Special Offers