missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL18BP Protein

A DNA sequence encoding the Homo sapiens (Human) IL18BP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8878-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016941

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001034748.1
IL18BP Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
10068
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL18BP
Recombinant Protein
E. coli
TPVSQTTTAA TASVRSTKDP CPSQPPVFPA AKQCPALEVT WPEVEVPLNG TLSLSCVACS RFPNFSILYW LGNGSFIEHL PGRLWEGSTS RERGSTGTQL CKALVLEQLT PALHSTNFSC VLVDPEQVVQ RHVVLAQLWA GLRATLPPTQ EAL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers