missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein
A DNA sequence encoding the Human IL-29 / Interleukin-29, was expressed in the hosts and tags indicated.
105.00€ - 1970.00€
Specifications
Accession Number | NP_742152.1 |
---|---|
Gene ID (Entrez) | 282618 |
Name | IL-29 / Interleukin-29 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953768
|
enQuireBio™
QP10410-EC-5UG |
5 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15933768
|
enQuireBio™
QP10410-EC-20UG |
20 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15923768
|
enQuireBio™
QP10410-EC-100UG |
100 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15943768
|
enQuireBio™
QP10410-EC-250UG |
250 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_742152.1 | |
IL-29 / Interleukin-29 Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPVPTSKPTT TGKGCHIGRF KSLSPQELAS FKKARDALEE SLKLKNWSCS SPVFPGNWDL RLLQVRERPV ALEAELALTL KVLEAAAGPA LEDVLDQPLH TLHHILSQLQ ACIQPQPTAG PRPRGRLHHW LHRLQEAPKK ESAGCLEASV TFNLFRLLTR DLKYVADGNL CLRTSTHPES T | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
282618 | |
Research Use Only | |
IFNL1 | |
The ED50 as determined by an anti-viral assay using human HepG2 cells infected with encephalomyocarditis is less than 5 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |