missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFIH1 Protein

A DNA sequence encoding the Homo sapiens (Human) IFIH1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8166-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033011

Quantity Price
1 3211.16€ / 1mg
3 3021.38€ / 1mg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_071451.2
IFIH1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
64135
Greater than 80% as determined by SDS-PAGE.
Research Use Only
IFIH1
Recombinant Protein
E. coli
KLTKLRNTIM EQYTRTEESA RGIIFTKTRQ SAYALSQWIT ENEKFAEVGV KAHHLIGAGH SSEFKPMTQN EQKEVISKFR TGKINLLIAT TVAEEGLDIK ECNIVIRYGL VTNEIAMVQA RGRARADEST YVLVAHSGSG VIEHETVNDF REKMMYKAIH CVQNMKPEEY AHKILELQMQ SIMEKKMKTK RNIAKHYKNN PSLITFLCKN CSVLACSGED IHVIEKMHHV NMTPEFKELY IVRENKALQK KCADYQINGE IICKCGQAWG TMMVHKGLDL PCLKIRNFVV VFKNNSTKKQ YKKWVELPIT FPNLDYSECC LFSDED
Videos
SDS
Documents
Certificates
Special Offers

Special Offers