missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IDH3B Protein

A DNA sequence encoding the Homo sapiens (Human) IDH3B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6193-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031460

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001317692.1
IDH3B Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
3420
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IDH3B
Recombinant Protein
E. coli
ASRSQAEDVR VEGSFPVTML PGDGVGPELM HAVKEVFKAA AVPVEFQEHH LSEVQNMASE EKLEQVLSSM KENKVAIIGK IHTPMEYKGE LASYDMRLRR KLDLFANVVH VKSLPGYMTR HNNLDLVIIR EQTEGEYSSL EHESARGVIE CLKIVTRAKS QRIAKFAFDY ATKKGRGKVT AVHKANIMKL GDGLFLQCCE EVAELYPKIK FETMIIDNCC MQLVQNPYQF DVLVMPNLYG NIIDNLAAGL VGGAGVVPGE SYSAEYAVFE TGARHPFAQA VGRNIANPTA MLLSASNMLR HLNLEYHSSM IADAVKKVIK VGKVRTRDMG GYSTTTDFIK SVIGHLQTKG S
Videos
SDS
Documents
Certificates
Special Offers

Special Offers