missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DMA Protein

A DNA sequence encoding the Homo sapiens (Human) HLA-DMA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7191-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027630

  • 2385.00€ / 1mg
Estimated Shipment: 20-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_006111.2
HLA-DMA Protein
Research Use Only
HLA-DMA
Recombinant Protein
E. coli
VPEAPTPMWP DDLQNHTFLH TVYCQDGSPS VGLSEAYDED QLFFFDFSQN TRVPRLPEFA DWAQEQGDAP AILFDKEFCE WMIQQIGPKL DGKIPVSRGF PIAEVFTLKP LEFGKPNTLV CFVSNLFPPM LTVNWQHHSV PVEGFGPTFV SAVDGLSFQA FSYLNFTPEP SDIFSCIVTH EIDRYTAIAY WVPRNALPSD LLENVLC
Greater than 90% as determined by SDS-PAGE.
3108
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers