missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone H2B type 1-N Protein

A DNA sequence encoding the Homo sapiens (Human) Histone H2B type 1-N, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8012-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022151

  • 2385.00€ / 1mg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_003511.1
Histone H2B type 1-N Protein
Research Use Only
HIST1H2BN
Recombinant Protein
E. coli
PEPSKSAPAP KKGSKKAVTK AQKKDGKKRK RSRKESYSVY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSSK
Greater than 90% as determined by SDS-PAGE.
8341
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers