missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNL3L Protein

A DNA sequence encoding the Homo sapiens (Human) GNL3L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8075-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022521

  • 530.00€ / 100µg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001171748.1
GNL3L Protein
Research Use Only
GNL3L
Recombinant Protein
E. coli
MMKLRHKNKK PGEGSKGHKK ISWPYPQPAK QNGKKATSKV PSAPHFVHPN DHANREAELK KKWVEEMREK QQAAREQERQ KRRTIESYCQ DVLRRQEEFE HKEEVLQELN MFPQLDDEAT RKAYYKEFRK VVEYSDVILE VLDARDPLGC RCFQMEEAVL RAQGNKKLVL VLNKIDLVPK EVVEKWLDYL RNELPTVAFK ASTQHQVKNL NRCSVPVDQA SESLLKSKAC FGAENLMRVL GNYCRLGEVR THIRVGVVGL PNVGKSSLIN SLKRSRACSV GAVPGITKFM QEVYLDKFIR LLDAPGIVPG PNSEVGTILR NCVHVQKLAD PVTPVETILQ RCNLEEISNY YGVSGFQTTE HFLTAVAHRL GKKKKGGLYS QEQAAKAVLA DWVSGKISFY IPPPATHTLP THLSAEIVKE MTEVFDIEDT EQANEDTMEC LATGESDELL GDTDPLEMEI KLLHSPMTKI ADAIENKTTV YKIGDLTGYC TNPNRHQMGW AKRNVDHRPK SNSMVDVCSV DRRSVLQRIM ETDPLQQGQA LASALKNKKK MQKRADKIAS KLSDSMMSAL DLSGNADDGV GD
Greater than 90% as determined by SDS-PAGE.
54552
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers