missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Galectin-1 / LGALS1 Protein

A DNA sequence encoding the Homo sapiens (Human) Galectin-1 / LGALS1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6294-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022250

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_002296.1
Galectin-1 / LGALS1 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3956
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LGALS1
Recombinant Protein
E. coli
ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers