missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human G-protein coupled receptor 161 Protein

A DNA sequence encoding the Homo sapiens (Human) G-protein coupled receptor 161, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8467-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024771

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001254538.1
G-protein coupled receptor 161 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
23432
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GPR161
Recombinant Protein
E. coli
MSLNSSLSCR KELSNLTEEE GGEGGVII
Videos
SDS
Documents
Certificates
Special Offers

Special Offers