missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-box only protein 1
A DNA sequence encoding the Homo sapiens (Human) F-box only protein 3, was expressed in the hosts and tags indicated.
202.00€ - 2765.00€
Specifications
Accession Number | NP_036307.2 |
---|---|
Gene ID (Entrez) | 26273 |
Name | F-box only protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029031
|
enQuireBio™
QP9935-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16049031
|
enQuireBio™
QP9935-YE-1MG |
1 mg |
2765.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16039031
|
enQuireBio™
QP9935-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16059031
|
enQuireBio™
QP9935-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16069031
|
enQuireBio™
QP9935-YE-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16079031
|
enQuireBio™
QP9935-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_036307.2 | |
F-box only protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
26273 | |
Research Use Only | |
FBXO3 | |
Recombinant Protein | |
Yeast | |
MAAMETETAP LTLESLPTDP LLLILSFLDY RDLINCCYVS RRLSQLSSHD PLWRRHCKKY WLISEEEKTQ KNQCWKSLFI DTYSDVGRYI DHYAAIKKAW DDLKKYLEPR CPRMVLSLKE GAREEDLDAV EAQIGCKLPD DYRCSYRIHN GQKLVVPGLL GSMALSNHYR SEDLLDVDTA AGGFQQRQGL KYCLPLTFCI HTGLSQYIAV EAAEGRNKNE VFYQCPDQMA RNPAAIDMFI IGATFTDWFT SYVKNVVSGG FPIIRDQIFR YVHDPECVAT TGDITVSVST SFLPELSSVH PPHYFFTYRI RIEMSKDALP EKACQLDSRY WRITNAKGDV EEVQGPGVVG EFPIISPGRV YEYTSCTTFS TTSGYMEGYY TFHFLYFKDK IFNVAIPRFH MACPTFRVSI ARLEMGPDEY EEMEEEEEEE EEEDEDDDSA DMDESDEDDE EERRRRVFDV PIRRRRCSRL F | |
Greater than 90% as determined by SDS-PAGE. |