missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ERVK-6 Protein
A DNA sequence encoding the Homo sapiens (Human) ERVK-6, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Name | ERVK-6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | ERVK-6 |
Protein Length | Extracellular Domain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040321
|
enQuireBio™
QP7708-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16060321
|
enQuireBio™
QP7708-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16050321
|
enQuireBio™
QP7708-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16070321
|
enQuireBio™
QP7708-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16080321
|
enQuireBio™
QP7708-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16090321
|
enQuireBio™
QP7708-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
ERVK-6 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
ERVK-6 | |
Recombinant Protein | |
E. coli | |
LPMPAGAAAA NYTYWAYVPF PPLIRAVTWM DNPTEVYVND SVWVPGPIDD RCPAKPEEEG MMINISIGYH YPPICLGRAP GCLMPAVQNW LVEVPTVSPI CRFTYHMVSG MSLRPRVNYL QDFSYQRSLK FRPKGKPCPK EIPKESKNTE VLVWEECVAN SAVILQNNEF GTIIDWAPRG QFYHNCSGQT QSCPSAQVSP AVDSDLTESL DKHKHKKLQS FYPWEWGEKG ISTPRPKIVS PVSGPEHPEL WRLTVASHHI RIWSGNQTLE TRDRKPFYTI DLNSSLTVPL QSCVKPPYML VVGNIVIKPD SQTITCENCR LLTCIDSTFN WQHRILLVRA REGVWIPVSM DRPWEASPSV HILTEVLKGV LNRSKRFIFT LIAVIMGLIA VTATAAVAGV ALHSSVQSVN FVNDWQKNST RLWNSQSSID QKLANQINDL RQTVIWMGDR LMSLEHRFQL QCDWNTSDFC ITPQIYNESE HHWDMVRRHL QGREDNLTLD ISKLKEQIFE ASKAHLNLVP GTEAIAGVAD GLANLNPVTW VKT | |
Greater than 90% as determined by SDS-PAGE. |