missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Epcr / PROCR Protein
A DNA sequence encoding the Homo sapiens (Human) Epcr / PROCR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8541-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006395.2 | |
Epcr / PROCR Protein | |
Research Use Only | |
PROCR | |
Recombinant Protein | |
E. coli | |
SQDASDGLQR LHMLQISYFR DPYHVWYQGN ASLGGHLTHV LEGPDTNTTI IQLQPLQEPE SWARTQSGLQ SYLLQFHGLV RLVHQERTLA FPLTIRCFLG CELPPEGSRA HVFFEVAVNG SSFVSFRPER ALWQADTQVT SGVVTFTLQQ LNAYNRTRYE LREFLEDTCV QYVQKHISAE NTKGSQTSRS YTS | |
Greater than 90% as determined by SDS-PAGE. |
10544 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |