missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dickkopf-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Dickkopf-related protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8081-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042551

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_055236.1
Dickkopf-related protein 1
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
27123
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DKK2
Recombinant Protein
E. coli
KLNSIKSSLG GETPGQAANR SAGMYQGLAF GGSKKGKNLG QAYPCSSDKE CEVGRYCHSP HQGSSACMVC RRKKKRCHRD GMCCPSTRCN NGICIPVTES ILTPHIPALD GTRHRDRNHG HYSNHDLGWQ NLGRPHTKMS HIKGHEGDPC LRSSDCIEGF CCARHFWTKI CKPVLHQGEV CTKQRKKGSH GLEIFQRCDC AKGLSCKVWK DATYSSKARL HVCQKI
Videos
SDS
Documents
Certificates
Special Offers

Special Offers