missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dickkopf-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Dickkopf-related protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8081-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022551

  • 530.00€ / 100µg
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_055236.1
Dickkopf-related protein 1
Research Use Only
DKK2
Recombinant Protein
E. coli
KLNSIKSSLG GETPGQAANR SAGMYQGLAF GGSKKGKNLG QAYPCSSDKE CEVGRYCHSP HQGSSACMVC RRKKKRCHRD GMCCPSTRCN NGICIPVTES ILTPHIPALD GTRHRDRNHG HYSNHDLGWQ NLGRPHTKMS HIKGHEGDPC LRSSDCIEGF CCARHFWTKI CKPVLHQGEV CTKQRKKGSH GLEIFQRCDC AKGLSCKVWK DATYSSKARL HVCQKI
Greater than 90% as determined by SDS-PAGE.
27123
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers