missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Death-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Death-associated protein 1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001278892.1 |
---|---|
Gene ID (Entrez) | 1611 |
Name | Death-associated protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034131
|
enQuireBio™
QP8344-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16054131
|
enQuireBio™
QP8344-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16044131
|
enQuireBio™
QP8344-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16064131
|
enQuireBio™
QP8344-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16074131
|
enQuireBio™
QP8344-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16084131
|
enQuireBio™
QP8344-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001278892.1 | |
Death-associated protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1611 | |
Research Use Only | |
DAP1 | |
Recombinant Protein | |
E. coli | |
SSPPEGKLET KAGHPPAVKA GGMRIVQKHP HTGDTKEEKD KDDQEWESPS PPKPTVFISG VIARGDKDFP PAAAQVAHQK PHASMDKHPS PRTQHIQQPR K | |
Greater than 90% as determined by SDS-PAGE. |