missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin 8 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 8, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001243222.1 |
---|---|
Gene ID (Entrez) | 3856 |
Name | Cytokeratin 8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16012180
|
enQuireBio™
QP6284-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16032180
|
enQuireBio™
QP6284-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16022180
|
enQuireBio™
QP6284-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16042180
|
enQuireBio™
QP6284-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052180
|
enQuireBio™
QP6284-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062180
|
enQuireBio™
QP6284-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001243222.1 | |
Cytokeratin 8 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3856 | |
Research Use Only | |
KRT8 | |
Recombinant Protein | |
E. coli | |
SIRVTQKSYK VSTSGPRAFS SRSYTSGPGS RISSSSFSRV GSSNFRGGLG GGYGGASGMG GITAVTVNQS LLSPLVLEVD PNIQAVRTQE KEQIKTLNNK FASFIDKVRF LEQQNKMLET KWSLLQQQKT ARSNMDNMFE SYINNLRRQL ETLGQEKLKL EAELGNMQGL VEDFKNKYED EINKRTEMEN EFVLIKKDVD EAYMNKVELE SRLEGLTDEI NFLRQLYEEE IRELQSQISD TSVVLSMDNS RSLDMDSIIA EVKAQYEDIA NRSRAEAESM YQIKYEELQS LAGKHGDDLR RTKTEISEMN RNISRLQAEI EGLKGQRASL EAAIADAEQR GELAIKDANA KLSELEAALQ RAKQDMARQL REYQELMNVK LALDIEIATY RKLLEGEESR LESGMQNMSI HTKTTSGYAG GLSSAYGGLT SPGLSYSLGS SFGSGAGSSS FSRTSSSRAV VVKKIETRDG KLVSESSDVL PK | |
Greater than 90% as determined by SDS-PAGE. |