missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 10, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_000412.3 |
---|---|
Gene ID (Entrez) | 3858 |
Name | Cytokeratin 10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16012120
|
enQuireBio™
QP6275-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16032120
|
enQuireBio™
QP6275-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16022120
|
enQuireBio™
QP6275-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16042120
|
enQuireBio™
QP6275-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052120
|
enQuireBio™
QP6275-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062120
|
enQuireBio™
QP6275-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Spezifikation
NP_000412.3 | |
Cytokeratin 10 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3858 | |
Research Use Only | |
KRT10 | |
Recombinant Protein | |
E. coli | |
MSVRYSSSKH YSSSRSGGGG GGGGCGGGGG VSSLRISSSK GSLGGGFSSG GFSGGSFSRG SSGGGCFGGS SGGYGGLGGF GGGSFRGSYG SSSFGGSYGG IFGGGSFGGG SFGGGSFGGG GFGGGGFGGG FGGGFGGDGG LLSGNEKVTM QNLNDRLASY LDKVRALEES NYELEGKIKE WYEKHGNSHQ GEPRDYSKYY KTIDDLKNQI LNLTTDNANI LLQIDNARLA ADDFRLKYEN EVALRQSVEA DINGLRRVLD ELTLTKADLE MQIESLTEEL AYLKKNHEEE MKDLRNVSTG DVNVEMNAAP GVDLTQLLNN MRSQYEQLAE QNRKDAEAWF NEKSKELTTE IDNNIEQISS YKSEITELRR NVQALEIELQ SQLALKQSLE ASLAETEGRY CVQLSQIQAQ ISALEEQLQQ IRAETECQNT EYQQLLDIKI RLENEIQTYR SLLEGEGSSG GGGRGGGSFG GGYGGGSSGG GSSGGGHGGG HGGSSGGGYG GGSSGGGSSG GGYGGGSSSG GHGGSSSGGY GGGSSGGGGG GYGGGSSGGG SSSGGGYGGG SSSGGHKSSS SGSVGESSSK GPRY | |
Greater than 90% as determined by SDS-PAGE. |