missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytokeratin 10 Protein

A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6275-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032120

  • 2385.00€ / 1mg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_000412.3
Cytokeratin 10 Protein
Research Use Only
KRT10
Recombinant Protein
E. coli
MSVRYSSSKH YSSSRSGGGG GGGGCGGGGG VSSLRISSSK GSLGGGFSSG GFSGGSFSRG SSGGGCFGGS SGGYGGLGGF GGGSFRGSYG SSSFGGSYGG IFGGGSFGGG SFGGGSFGGG GFGGGGFGGG FGGGFGGDGG LLSGNEKVTM QNLNDRLASY LDKVRALEES NYELEGKIKE WYEKHGNSHQ GEPRDYSKYY KTIDDLKNQI LNLTTDNANI LLQIDNARLA ADDFRLKYEN EVALRQSVEA DINGLRRVLD ELTLTKADLE MQIESLTEEL AYLKKNHEEE MKDLRNVSTG DVNVEMNAAP GVDLTQLLNN MRSQYEQLAE QNRKDAEAWF NEKSKELTTE IDNNIEQISS YKSEITELRR NVQALEIELQ SQLALKQSLE ASLAETEGRY CVQLSQIQAQ ISALEEQLQQ IRAETECQNT EYQQLLDIKI RLENEIQTYR SLLEGEGSSG GGGRGGGSFG GGYGGGSSGG GSSGGGHGGG HGGSSGGGYG GGSSGGGSSG GGYGGGSSSG GHGGSSSGGY GGGSSGGGGG GYGGGSSGGG SSSGGGYGGG SSSGGHKSSS SGSVGESSSK GPRY
Greater than 90% as determined by SDS-PAGE.
3858
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers